Learn More
Sino Biological VCAM1/VCAM-1/CD106 Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10113MM11200
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human VCAM1/VCAM-1/CD106 (rh VCAM1/VCAM-1/CD106; Catalog#10113-H08H; NP_001069.1; Met1-Pro697). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-1167