Learn More
Sino Biological VCAM1/VCAM-1/CD106 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10113T5650
Produced in rabbits immunized with purified, recombinant Human VCAM1/VCAM-1/CD106 (rh VCAM1/VCAM-1/CD106; Catalog#10113-H08H; NP_001069.1; Met1-Pro697). VCAM1/VCAM-1/CD106 specific IgG was purified by Human VCAM1/VCAM-1/CD106 affinity chromatography.
Catalog No. 50-172-1183