Learn More
Sino Biological DSG1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101147T3250
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human DSG1, and purified by antigen affinity chromatography.
Catalog No. 50-172-1192