Learn More
Sino Biological CD146 / MCAM Antibody, Rabbit PAb DFS Item
Manufacturer:  Sino Biological 10115RP01200
Produced in rabbits immunized with purified, recombinant Human CD146 / MCAM extracellular domain (rhCD146; Catalog#10115-H08H; NP_006491.2; Met 1-Gly 559). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-1248