Learn More
Sino Biological Aryl Hydrocarbon Receptor Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101180T42100
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Aryl Hydrocarbon Receptor, and purified by antigen affinity chromatography.
Catalog No. 50-172-1283