Learn More
Sino Biological HMGB3 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101181T08200
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the human HMGB3, and purified by antigen affinity chromatography.
Catalog No. 50-172-1287