Learn More
Sino Biological TLE1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101194T4250
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human TLE1, and purified by antigen affinity chromatography.
Catalog No. 50-172-1315