Learn More
Sino Biological IKBKB / IKK beta Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101200T4050
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human IKBKB / IKK beta, and purified by antigen affinity chromatography.
Catalog No. 50-172-1336