Learn More
Sino Biological Human IL-1RAcP / IL-1R3 Protein (His & Fc Tag) DFS Item
Manufacturer:  Sino Biological 10121H03H200
A DNA sequence encoding the human IL1R3 (NP_002173.1) extracellular domain (Met 1-Glu 359) was fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4062