Learn More
Sino Biological Human IL-1RAcP / IL-1R3 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10121H08H200
A DNA sequence encoding the human IL1R3 (NP_002173.1) extracellular domain (Met 1-Glu 359) was expressed, fused with the a C-terminal polyhistidine tag.
Catalog No. 50-161-4064