Learn More
Sino Biological IL-1RAcP / IL-1R3 Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10121MM05200
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL1R3 / IL1RAP (rh IL1R3; Catalog#10121-H08H; NP_002173.1; Met 1-Glu 359). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-1388