Learn More
Sino Biological IL-1RAcP / IL-1R3 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10121T4850
Produced in rabbits immunized with purified, recombinant Human IL-1RAcP / IL-1R3 (rh IL-1RAcP / IL-1R3; Catalog#10121-H08H; NP_002173.1; Met1-Glu359). IL-1RAcP / IL-1R3 specific IgG was purified by Human IL-1RAcP / IL-1R3 affinity chromatography.
Catalog No. 50-172-1404