Learn More
Sino Biological IL1F10 / IL-38 Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10122MM06200
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL1F10 / IL-38 (rh IL1F10 / IL-38; Catalog#10122-H08Y; AAK68048.1; Met1-Trp152). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-1409