Learn More
Sino Biological PAK1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101224T3850
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Rat PAK1, and purified by antigen affinity chromatography.
Catalog No. 50-172-1407