Learn More
Sino Biological Human IL-1RA / IL1RN Protein DFS Item
Manufacturer:  Sino Biological 10123HNAE100
A DNA sequence encoding the mature form of human IL1RA isoform1 (NP_776214.1) (Arg 26-Glu 177) was expressed.
Catalog No. 50-161-4069