Learn More
Sino Biological IL-1RA / IL1RN Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10123RP02100
Produced in rabbits immunized with purified, recombinant Human IL-1RA (rh IL-1RA; Catalog#10123-HNAE; NP_776214.1; Arg26-Glu177). IL-1RA specific IgG was purified by Human IL-1RA affinity chromatography.
Catalog No. 50-172-1457