Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Sino Biological VDAC1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101236T44100
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human VDAC1, and purified by antigen affinity chromatography.
Catalog No. 50-172-1426