Learn More
Sino Biological IL36G Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10124MM10200
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL36G (rh IL36G; Catalog#10124-HNAE; Q9NZH8-1; Met1-Asp169). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-1488