Learn More
Sino Biological IL36G / IL1F9 Antibody, Rabbit MAb DFS Item
Manufacturer:  Sino Biological 10124R01350
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL36G / IL1F9 (rh IL36G / IL1F9; Catalog#10124-H07E1; NP_062564; Ser18-Asp169).
Catalog No. 50-172-1491