Learn More
Sino Biological H2AFM / HIST1H2AB Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101245T3250
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human H2AFM / HIST1H2AB, and purified by antigen affinity chromatography.
Catalog No. 50-172-1477