Learn More
Sino Biological LINGO1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101252T08200
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human LINGO1, and purified by antigen affinity chromatography.
Catalog No. 50-172-1502