Learn More
Sino Biological Human IL1R1 / CD121a Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10126H02H50
A DNA sequence encoding the extracellular domain of human IL1R1 (NP_000868.1) (Met 1-Thr 332) was fused with human IgG1 Fc region at the C-terminus.
Catalog No. 50-161-4080