Learn More
Sino Biological IL-1 alpha / IL-1F1 / IL1A Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10128T16200
Produced in rabbits immunized with purified, recombinant Human IL-1 alpha / IL-1F1 / IL1A (rh IL-1 alpha / IL-1F1 / IL1A; Catalog#10128-HNCH; NP_000566.3; Ser113-Ala271). IL-1 alpha / IL-1F1 / IL1A specific IgG was purified by Human IL-1 alpha / IL-1F1 / IL1A affinity chromatography.
Catalog No. 50-172-1581