Learn More
Sino Biological NANOG / Nanog homeobox Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101287T0850
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human NANOG / Nanog homeobox, and purified by antigen affinity chromatography.
Catalog No. 50-172-1565