Learn More
Sino Biological Human PCSK1 / NEC1 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10129H08H10
A DNA sequence encoding the human PCSK1 (NP_000430.3) (Met 1-Arg 617) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4094