Learn More
Sino Biological Human PIGR Protein (365 Ser/Gly, His Tag) DFS Item
Manufacturer:  Sino Biological 10131H08H50
A DNA sequence encoding the extracellular domain (Met 1-Arg 638, 365 Ser/Gly) of human PIGR (NP_002635.2) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4099