Learn More
Sino Biological PIGR / Poly-Ig receptor Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10131RP02200
Produced in rabbits immunized with purified, recombinant Human PIGR extracellular domain (rh PIGR; Catalog#10131-H08H; NP_002635.2; Met 1-Arg 638). PIGR specific IgG was purified by human PIGR affinity chromatography.
Catalog No. 50-172-1638