Learn More
Sino Biological Human CCL2 / MCP-1 / MCP1 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10134H08Y50
A DNA sequence encoding the human CCL2 (NP_002973.1) (Gln24-Thr99) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4103