Learn More
Sino Biological MCP-1 / CCL2 Antibody, Rabbit PAb DFS Item
Manufacturer:  Sino Biological 10134RP01100
Produced in rabbits immunized with purified, recombinant Human MCP-1 / CCL2 (rh MCP-1 / CCL2; Catalog#10134-H08Y; NP_002973.1; Gln24-Thr99). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-1701