Learn More
Sino Biological MCP-1/CCL2 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10134T2450
Produced in rabbits immunized with purified, recombinant Human MCP-1/CCL2 (rh MCP-1/CCL2; Catalog#10134-H08Y; NP_002973.1; Gln24-Thr99). MCP-1/CCL2 specific IgG was purified by Human MCP-1/CCL2 affinity chromatography.
Catalog No. 50-172-1709