Learn More
Sino Biological 40S ribosomal protein S3 / RPS3 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101346T3650
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human 40S ribosomal protein S3/RPS3, and purified by antigen affinity chromatography.
Catalog No. 50-172-1679