Learn More
Sino Biological Human VEGFR1 / FLT-1 Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10136H02H50
A DNA sequence encoding the human FLT1 (NP_001153503.1) (Met1-Ile328) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-4105