Learn More
Sino Biological Human VEGFR1 / FLT-1 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10136H08H150
A DNA sequence encoding the human FLT1 (NP_002010.2) (Met1-Ile328) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-4108