Learn More
Sino Biological VEGFR1 / FLT-1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10136RP02200
Produced in rabbits immunized with purified, recombinant Human VEGFR1 / FLT-1 (rh VEGFR1 / FLT-1; Catalog#10136-H08H; P17948-1; Met 1-Asn 756). VEGFR1 / FLT-1 specific IgG was purified by Human VEGFR1 / FLT-1 affinity chromatography.
Catalog No. 50-172-1750