Learn More
Sino Biological Fas Ligand / FASLG / CD95L Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101369T36100
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Fas Ligand / FASLG / CD95L, and purified by antigen affinity chromatography.
Catalog No. 50-172-1734