Learn More
Sino Biological Human IL-1 beta / IL1B Protein DFS Item
Manufacturer:  Sino Biological 10139HNAE20
A DNA sequence encoding the mature form of human IL1β (NP_000567.1) (Ala 117-Ser 269) was expressed.
Catalog No. 50-161-4114