Learn More
Sino Biological IL1B / IL-1B / IL-1 beta Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10139MM09100
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL1B / IL-1B / IL-1 beta (rh IL1B / IL-1B / IL-1 beta; Catalog#10139-H07E; NP_000567.1; Met1-Ser269). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-1784