Learn More
Sino Biological MAP2 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101399T08200
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human MAP2, and purified by antigen affinity chromatography.
Catalog No. 50-172-1774