Learn More
Sino Biological CD171 / N-CAML1 / L1CAM Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10140RP02100
Produced in rabbits immunized with purified, recombinant Human CD171 (rh CD171; Catalog#10140-H08H; NP_000416.1; Met 1-Glu 1120). CD171 specific IgG was purified by human CD171 affinity chromatography .
Catalog No. 50-172-1831