Learn More
Sino Biological Human SerpinC1 / AntithrombinIII / ATIII Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10142H08H20
A DNA sequence encoding the human SerpinC1 (NP_000479.1) (Met 1-Lys 464) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4120