Learn More
Sino Biological TCF7L1 / TCF3 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101427T3650
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human TCF7L1 / TCF3, and purified by antigen affinity chromatography.
Catalog No. 50-172-1845