Learn More
Sino Biological CHL1 / LICAM2 / CALL Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10143MM051
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CHL1 / LICAM2 / CALL (rh CHL1 / LICAM2 / CALL; Catalog#10143-H08H; AAI04919.1; Met1-Gln1080). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-1870