Learn More
Sino Biological Human Cadherin-8 / CDH8 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10144H08H10
A DNA sequence encoding the extracellular domain of human CDH8 (NP_001787.2) (Met 1-Met 621) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-4123