Learn More
Sino Biological Cadherin-8 / CDH8 Antibody, Rabbit PAb DFS Item
Manufacturer:  Sino Biological 10144RP01400
Produced in rabbits immunized with purified, recombinant Human Cadherin-8 / CDH8 (rh Cadherin-8 / CDH8; Catalog#10144-H08H; NP_001787.2; Met1-Met621). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-172-1884