Learn More
Sino Biological Cadherin-8 / CDH8 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10144T16200
Produced in rabbits immunized with purified, recombinant Human Cadherin-8 / CDH8 (rh Cadherin-8 / CDH8; Catalog#10144-H08H; NP_001787.2; Met1-Met621). Cadherin-8 / CDH8 specific IgG was purified by Human Cadherin-8 / CDH8 affinity chromatography.
Catalog No. 50-172-1886