Learn More
Sino Biological TLR4 / TLR-4 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10146T16200
Produced in rabbits immunized with purified, recombinant Human TLR4 / TLR-4 (rh TLR4 / TLR-4; Catalog#10146-H08B; O00206-1; Met1-Lys631). TLR4 / TLR-4 specific IgG was purified by Human TLR4 / TLR-4 affinity chromatography.
Catalog No. 50-172-1919