Learn More
Sino Biological E6AP Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101460T1050
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human E6AP, and purified by antigen affinity chromatography.
Catalog No. 50-172-1899