Learn More
Sino Biological VEGF/VEGFA Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 101465MM13200
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with a synthetic peptide corresponding to the C-terminus of the Human VEGF/VEGFA. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-172-1913