Learn More
Sino Biological WDR77 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 101467T4250
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human WDR77, and purified by antigen affinity chromatography.
Catalog No. 50-172-1917