Learn More
Sino Biological Human RSK4 / RPS6KA6 Protein (GST Tag) DFS Item
Manufacturer:  Sino Biological 10147H09B50
A DNA sequence encoding the full length of human RSK4 (NP_055311.1) (Met 1-Leu 745) was fused with the GST tag at the N-terminus.
Catalog No. 50-161-4129